MilliporeSigma
  • Calsyntenin-1 regulates targeting of dendritic NMDA receptors and dendritic spine maturation in CA1 hippocampal pyramidal cells during postnatal development.
Calsyntenin-1 regulates targeting of dendritic NMDA receptors and dendritic spine maturation in CA1 hippocampal pyramidal cells during postnatal development.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-06-27)
Jeanne Ster, Martin Steuble, Clara Orlando, Tu-My Diep, Alexander Akhmedov, Olivier Raineteau, Vincent Pernet, Peter Sonderegger, Urs Gerber
ABSTRACT
Calsyntenin-1 is a transmembrane cargo-docking protein important for kinesin-1-mediated fast transport of membrane-bound organelles that exhibits peak expression levels at postnatal day 7. However, its neuronal function during postnatal development remains unknown. We generated a knock-out mouse to characterize calsyntenin-1 function in juvenile mice. In the absence of calsyntenin-1, synaptic transmission was depressed. To address the mechanism, evoked EPSPs were analyzed revealing a greater proportion of synaptic GluN2B subunit-containing receptors typical for less mature synapses. This imbalance was due to a disruption in calsyntenin-1-mediated dendritic transport of NMDA receptor subunits. As a consequence of increased expression of GluN2B subunits, NMDA receptor-dependent LTP was enhanced at Schaffer collateral-CA1 pyramidal cell synapses. Interestingly, these defects were accompanied by a decrease in dendritic arborization and increased proportions of immature filopodia-like dendritic protrusions at the expense of thin-type dendritic spines in CA1 pyramidal cells. Thus, these results highlight a key role for calsyntenin-1 in the transport of NMDA receptors to synaptic targets, which is necessary for the maturation of neuronal circuits during early development.
Sigma-Aldrich
Anti-NMDAR2A Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse
Sigma-Aldrich
Anti-NMDAR2B Antibody, pain, serum, Chemicon®