MilliporeSigma
  • Endothelial LRP1 transports amyloid-β(1-42) across the blood-brain barrier.
Endothelial LRP1 transports amyloid-β(1-42) across the blood-brain barrier.
The Journal of clinical investigation (2015-12-01)
Steffen E Storck, Sabrina Meister, Julius Nahrath, Julius N Meißner, Nils Schubert, Alessandro Di Spiezio, Sandra Baches, Roosmarijn E Vandenbroucke, Yvonne Bouter, Ingrid Prikulis, Carsten Korth, Sascha Weggen, Axel Heimann, Markus Schwaninger, Thomas A Bayer, Claus U Pietrzik
ABSTRACT
According to the neurovascular hypothesis, impairment of low-density lipoprotein receptor-related protein-1 (LRP1) in brain capillaries of the blood-brain barrier (BBB) contributes to neurotoxic amyloid-β (Aβ) brain accumulation and drives Alzheimer's disease (AD) pathology. However, due to conflicting reports on the involvement of LRP1 in Aβ transport and the expression of LRP1 in brain endothelium, the role of LRP1 at the BBB is uncertain. As global Lrp1 deletion in mice is lethal, appropriate models to study the function of LRP1 are lacking. Moreover, the relevance of systemic Aβ clearance to AD pathology remains unclear, as no BBB-specific knockout models have been available. Here, we developed transgenic mouse strains that allow for tamoxifen-inducible deletion of Lrp1 specifically within brain endothelial cells (Slco1c1-CreER(T2) Lrp1(fl/fl) mice) and used these mice to accurately evaluate LRP1-mediated Aβ BBB clearance in vivo. Selective deletion of Lrp1 in the brain endothelium of C57BL/6 mice strongly reduced brain efflux of injected [125I] Aβ(1-42). Additionally, in the 5xFAD mouse model of AD, brain endothelial-specific Lrp1 deletion reduced plasma Aβ levels and elevated soluble brain Aβ, leading to aggravated spatial learning and memory deficits, thus emphasizing the importance of systemic Aβ elimination via the BBB. Together, our results suggest that receptor-mediated Aβ BBB clearance may be a potential target for treatment and prevention of Aβ brain accumulation in AD.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Mouse IgG (H+L) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution