MilliporeSigma
  • Generation and characterization of a novel recombinant antibody against LMP1-TES1 of Epstein-Barr virus isolated by phage display.
Generation and characterization of a novel recombinant antibody against LMP1-TES1 of Epstein-Barr virus isolated by phage display.
Viruses (2013-04-24)
Dawei Zhang, Yuan Mao, Qing Cao, Lin Xiong, Juan Wen, Renjie Chen, Jin Zhu
ABSTRACT
Latent Membrane Protein 1 (LMP1) is a primary target for controlling tumorigenesis in Epstein-Barr virus related malignancies; in this study, we aimed to develop a specific antibody against the TES1 domain of the oncogenic LMP1. We screened a full human naïve Fab phage library against TES1 peptide, which consisted of C terminal-activating regions proximal 44 amino acids. After three rounds of panning, enrichment and testing by phage ELISA and further analyzed by DNA sequencing, we selected a phage clone with the highest affinity to LMP1-TES1 and designated it as htesFab. The positive clone was expressed in Escherichia coli and the purified htesFab was characterized for its binding specificity and affinity to LMP1. ELISA, immunofluorescence and FACS analysis confirmed that htesFab could recognize LMP1 TES1 both in vitro and in LMP1 expressing HNE2-LMP1 cells. Furthermore, MTT assay showed that htesFab inhibited the proliferation of HNE2-LMP1 cells in a dose-dependent manner. In summary, this study reported the isolation and characterization of human Fab, which specifically targets the C terminal region/TES1 of LMP1, and has potential to be developed as novel tool for the diagnosis and therapy of Epstein-Barr virus related carcinoma.
Sigma-Aldrich
Anti-Human IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution