MilliporeSigma
  • Chronic epithelial NF-κB activation accelerates APC loss and intestinal tumor initiation through iNOS up-regulation.
Chronic epithelial NF-κB activation accelerates APC loss and intestinal tumor initiation through iNOS up-regulation.
Proceedings of the National Academy of Sciences of the United States of America (2012-08-16)
Helena Shaked, Lorne J Hofseth, Alena Chumanevich, Alexander A Chumanevich, Jin Wang, Yinsheng Wang, Koji Taniguchi, Monica Guma, Steve Shenouda, Hans Clevers, Curtis C Harris, Michael Karin
ABSTRACT
The role of NF-κB activation in tumor initiation has not been thoroughly investigated. We generated Ikkβ(EE)(IEC) transgenic mice expressing constitutively active IκB kinase β (IKKβ) in intestinal epithelial cells (IECs). Despite absence of destructive colonic inflammation, Ikkβ(EE)(IEC) mice developed intestinal tumors after a long latency. However, when crossed to mice with IEC-specific allelic deletion of the adenomatous polyposis coli (Apc) tumor suppressor locus, Ikkβ(EE)(IEC) mice exhibited more β-catenin(+) early lesions and visible small intestinal and colonic tumors relative to Apc(+/ΔIEC) mice, and their survival was severely compromised. IEC of Ikkβ(EE)(IEC) mice expressed high amounts of inducible nitric oxide synthase (iNOS) and elevated DNA damage markers and contained more oxidative DNA lesions. Treatment of Ikkβ(EE)(IEC)/Apc(+/ΔIEC) mice with an iNOS inhibitor decreased DNA damage markers and reduced early β-catenin(+) lesions and tumor load. The results suggest that persistent NF-κB activation in IEC may accelerate loss of heterozygocity by enhancing nitrosative DNA damage.
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Nitrotyrosine antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution