MilliporeSigma
JARID1B is a luminal lineage-driving oncogene in breast cancer.
Cancer cell (2014-06-18)
Shoji Yamamoto, Zhenhua Wu, Hege G Russnes, Shinji Takagi, Guillermo Peluffo, Charles Vaske, Xi Zhao, Hans Kristian Moen Vollan, Reo Maruyama, Muhammad B Ekram, Hanfei Sun, Jee Hyun Kim, Kristopher Carver, Mattia Zucca, Jianxing Feng, Vanessa Almendro, Marina Bessarabova, Oscar M Rueda, Yuri Nikolsky, Carlos Caldas, X Shirley Liu, Kornelia Polyak
ABSTRACT
Recurrent mutations in histone-modifying enzymes imply key roles in tumorigenesis, yet their functional relevance is largely unknown. Here, we show that JARID1B, encoding a histone H3 lysine 4 (H3K4) demethylase, is frequently amplified and overexpressed in luminal breast tumors and a somatic mutation in a basal-like breast cancer results in the gain of unique chromatin binding and luminal expression and splicing patterns. Downregulation of JARID1B in luminal cells induces basal genes expression and growth arrest, which is rescued by TGFβ pathway inhibitors. Integrated JARID1B chromatin binding, H3K4 methylation, and expression profiles suggest a key function for JARID1B in luminal cell-specific expression programs. High luminal JARID1B activity is associated with poor outcome in patients with hormone receptor-positive breast tumors.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-KDM5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution