MilliporeSigma
  • Coupling an EML4-ALK-centric interactome with RNA interference identifies sensitizers to ALK inhibitors.
Coupling an EML4-ALK-centric interactome with RNA interference identifies sensitizers to ALK inhibitors.
Science signaling (2016-11-05)
Guolin Zhang, Hannah Scarborough, Jihye Kim, Andrii I Rozhok, Yian Ann Chen, Xiaohui Zhang, Lanxi Song, Yun Bai, Bin Fang, Richard Z Liu, John Koomen, Aik Choon Tan, James Degregori, Eric B Haura
ABSTRACT
Patients with lung cancers harboring anaplastic lymphoma kinase (ALK) gene fusions benefit from treatment with ALK inhibitors, but acquired resistance inevitably arises. A better understanding of proximal ALK signaling mechanisms may identify sensitizers to ALK inhibitors that disrupt the balance between prosurvival and proapoptotic effector signals. Using affinity purification coupled with mass spectrometry in an ALK fusion lung cancer cell line (H3122), we generated an ALK signaling network and investigated signaling activity using tyrosine phosphoproteomics. We identified a network of 464 proteins composed of subnetworks with differential response to ALK inhibitors. A small hairpin RNA screen targeting 407 proteins in this network revealed 64 and 9 proteins that when knocked down sensitized cells to crizotinib and alectinib, respectively. Among these, knocking down fibroblast growth factor receptor substrate 2 (FRS2) or coiled-coil and C2 domain-containing protein 1A (CC2D1A), both scaffolding proteins, sensitized multiple ALK fusion cell lines to the ALK inhibitors crizotinib and alectinib. Collectively, our data set provides a resource that enhances our understanding of signaling and drug resistance networks consequent to ALK fusions and identifies potential targets to improve the efficacy of ALK inhibitors in patients.
Sigma-Aldrich
Anti-CC2D1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)