MilliporeSigma
OGT restrains the expansion of DNA damage signaling.
Nucleic acids research (2016-11-02)
Qiang Chen, Xiaochun Yu
ABSTRACT
O-linked N-acetylglucosamine linkage (O-GlcNAcylation) to serine or threonine residues regulates numerous biological processes; however, its role in DNA damage response remains elusive. Here, we found that O-GlcNAcylation is induced by DNA damage response. O-GlcNAc transferase (OGT), the solo enzyme for O-GlcNAcylation, relocates to the sites of DNA damage and induces the O-GlcNAcylation of histone H2AX and mediator of DNA damage checkpoint 1 (MDC1). The O-GlcNAcylation negatively regulates DNA double-strand break-induced phosphorylation of H2AX and MDC1 by restraining the expansion of these phosphorylation events from the sites of DNA damage. Therefore, our study reveals the molecular mechanism and biological function of OGT-dependent O-GlcNAcylation in response to DNA damage.
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Anti-MDC1 antibody, Mouse monoclonal, clone MDC1-50, purified from hybridoma cell culture