MilliporeSigma
Optical Dedoping Mechanism for P3HT:F4TCNQ Mixtures.
The journal of physical chemistry letters (2016-11-04)
Jack Fuzell, Ian E Jacobs, Sophia Ackling, Thomas F Harrelson, David M Huang, Delmar Larsen, Adam J Moulé
ABSTRACT
Doping-induced solubility control (DISC) is a recently introduced photolithographic technique for semiconducting polymers, which utilizes reversible changes in polymer solubility upon doping to allow the polymer to function as its own photoresist. Central to this process is a wavelength sensitive optical dedoping reaction, which is poorly understood but generates subdiffraction-limited topographic features and provides optical control of the polymer doping level. Here, we examine the mechanism of optical dedoping in the semiconducting polymer poly-3-hexylthiophene (P3HT) doped by 2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane (F4TCNQ), via a combination of ultrafast and steady-state spectroscopy, ab initio calculations, and multidimensional NMR. A simple photoinduced back electron transfer mechanism from reduced F4TCNQ to oxidized P3HT does not explain the observed photophysics. Instead, photoexcited F4TCNQ* reacts with THF solvent molecules to form a neutral, nondoping, and highly soluble F4TCNQ-THF complex. Hence, ionized F4TCNQ is removed from the P3HT indirectly by depletion of the neutral F4TCNQ. Because the reaction involves only the dopant and similar photoreactivity would expected for most other dopant molecules, we expect optical DISC patterning should be generalizable to a wide range of polymer:dopant systems.
Sigma-Aldrich
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 97%