MilliporeSigma
  • Enzymatic characterization of recombinant rat DDHD2: a soluble diacylglycerol lipase.
Enzymatic characterization of recombinant rat DDHD2: a soluble diacylglycerol lipase.
Journal of biochemistry (2016-11-02)
Mari Araki, Noriyasu Ohshima, Chizu Aso, Akimitsu Konishi, Hideru Obinata, Kazuaki Tatei, Takashi Izumi
ABSTRACT
DDHD2 has been reported to exhibit phospholipase A
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium glycocholate hydrate, ≥95% (TLC)