MilliporeSigma
  • Effects of BMS-986094, a Guanosine Nucleotide Analogue, on Mitochondrial DNA Synthesis and Function.
Effects of BMS-986094, a Guanosine Nucleotide Analogue, on Mitochondrial DNA Synthesis and Function.
Toxicological sciences : an official journal of the Society of Toxicology (2016-07-29)
Bethany R Baumgart, Faye Wang, Jae Kwagh, Chris Storck, Catherine Euler, Megan Fuller, Damir Simic, Suresh Sharma, Jamie J Arnold, Craig E Cameron, Terry R Van Vleet, Oliver Flint, Roderick T Bunch, Marc H Davies, Michael J Graziano, Thomas P Sanderson
ABSTRACT
BMS-986094, the prodrug of a guanosine nucleotide analogue (2'-C-methylguanosine), was withdrawn from clinical trials due to serious safety issues. Nonclinical investigative studies were conducted as a follow up to evaluate the potential for BMS-986094-related mitochondrial-toxicity. In vitro, BMS-986094 was applied to human hepatoma cells (HepG2 and Huh-7) or cardiomyocytes (hiPSCM) up to 19 days to assess mitochondrial DNA content and specific gene expression. There were no mitochondrial DNA changes at concentrations ≤10 µM. Transcriptional effects, such as reductions in Huh-7 MT-ND1 and MT-ND5 mRNA content and hiPSCM MT-ND1, MT-COXII, and POLRMT protein expression levels, occurred only at cytotoxic concentrations (≥10 µM) suggesting these transcriptional effects were a consequence of the observed toxicity. Additionally, BMS-986094 has a selective weak affinity for inhibition of RNA polymerases as opposed to DNA polymerases. In vivo, BMS-986094 was given orally to cynomolgus monkeys for 3 weeks or 1 month at doses of 15 or 30 mg/kg/day. Samples of heart and kidney were collected for assessment of mitochondrial respiration, mitochondrial DNA content, and levels of high energy substrates. Although pronounced cardiac and renal toxicities were observed in some monkeys at 30 mg/kg/day treated for 3-4 weeks, there were no changes in mitochondrial DNA content or ATP/GTP levels. Collectively, these data suggest that BMS-986094 is not a direct mitochondrial toxicant.
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%
Sigma-Aldrich
BMS-986094, ≥98% (HPLC)