MilliporeSigma
  • Downregulation of Semaphorin-3F is associated with poor prognostic significance in osteosarcoma patients.
Downregulation of Semaphorin-3F is associated with poor prognostic significance in osteosarcoma patients.
American journal of cancer research (2016-11-09)
Ming-Han Liu, Wen-Juan Fu, You-Hong Cui, Qiao-Nan Guo, Yue Zhou
ABSTRACT
The secreted axonal guidance molecular, Semaphorin-3F (SEMA3F), is widely expressed outside the central nervous system and inhibits tumor growth and metastasis. However, the role of SEMA3F expression in the osteosarcoma prognosis has not been elaborated. This study aimed to evaluate SEMA3F expression level in osteosarcoma and assess its prognostic value for patients. SEMA3F protein expression was detected by immunohistochemistry (IHC) in 112 cases of osteosarcoma. Kaplan-Meier analysis and Cox regression analysis were performed to evaluate the prognostic significance of SEMA3F. The results showed that the overall survival and metastasis-free survival of patients with negative SEMA3F expression were significantly shorter than patients with positive expression (both
Sigma-Aldrich
MISSION® esiRNA, targeting human SEMA3F
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder