MilliporeSigma
  • Thermodynamics of biphasic lanthanide extraction by tripodal diglycolamide: a solution calorimetry study.
Thermodynamics of biphasic lanthanide extraction by tripodal diglycolamide: a solution calorimetry study.
Dalton transactions (Cambridge, England : 2003) (2016-11-02)
Seraj A Ansari, Prasanta K Mohapatra, Willem Verboom, Linfeng Rao
ABSTRACT
Isothermal titration calorimetry was employed for the direct measurement of the enthalpy of extraction (ΔH