MilliporeSigma
The Diphosphorus Complex [Cp
European journal of inorganic chemistry (2016-11-22)
Mehdi Elsayed Moussa, Bianca Attenberger, Martin Fleischmann, Andrea Schreiner, Manfred Scheer
ABSTRACT
The three-component reaction of the tetrahedral diphosphorus complex [Cp
Sigma-Aldrich
2,2′-Bipyrimidine, 95%