MilliporeSigma
  • Comparison of the pharmacokinetics between L-BPA and L-FBPA using the same administration dose and protocol: a validation study for the theranostic approach using [
Comparison of the pharmacokinetics between L-BPA and L-FBPA using the same administration dose and protocol: a validation study for the theranostic approach using [
BMC cancer (2016-11-09)
Tsubasa Watanabe, Yoshihide Hattori, Youichiro Ohta, Miki Ishimura, Yosuke Nakagawa, Yu Sanada, Hiroki Tanaka, Satoshi Fukutani, Shin-Ichiro Masunaga, Masahiro Hiraoka, Koji Ono, Minoru Suzuki, Mitsunori Kirihata
ABSTRACT
Boron neutron capture therapy (BNCT) is a cellular-level particle radiation therapy that combines the selective delivery of boron compounds to tumour tissue with neutron irradiation. L-p-Boronophenylalanine (L-BPA) is a boron compound now widely used in clinical situations. Determination of the boron distribution is required for successful BNCT prior to neutron irradiation. Thus, positron emission tomography with [ SCC-VII was subcutaneously inoculated into the legs of C3H/He mice. The same dose of L-BPA or [ There were no differences between L-BPA and [ No difference was noted in the time course of the boron concentrations in tumour tissue and normal tissues between L-BPA and [
Sigma-Aldrich
4-Borono-L-phenylalanine, ≥95.0% (HPLC)