MilliporeSigma
  • Inhibition of Glutaminolysis Inhibits Cell Growth via Down-regulating Mtorc1 Signaling in Lung Squamous Cell Carcinoma.
Inhibition of Glutaminolysis Inhibits Cell Growth via Down-regulating Mtorc1 Signaling in Lung Squamous Cell Carcinoma.
Anticancer research (2016-10-30)
Xulu Ye, Qiliang Zhou, Yoshifumi Matsumoto, Masato Moriyama, Shun Kageyama, Masaaki Komatsu, Seijiro Satoh, Masanori Tsuchida, Yasuo Saijo
ABSTRACT
Inhibition of glutaminolysis has been reported as a promising therapeutic strategy to target several solid carcinomas. We aimed to investigate the effects of glutaminolysis on cell proliferation in lung squamous cell carcinoma cell lines and to explore the potential of targeting glutaminolysis as an anticancer strategy. Glutamine (Gln) dependence was assessed in six lung squamous cell carcinoma cell lines. Cell proliferation, mammalian target of rapamycin complex 1 (mTORC1) activity and the induction of autophagy were assessed after inhibition of glutaminolysis via Gln depletion or glutaminase (GLS) inhibition. Five of six lung squamous cell carcinoma cell lines exhibited glutamine-dependence. The extent of dependence was correlated with the mRNA levels of GLS1/GLS2. Inhibition of glutaminolysis inhibited cell proliferation by down-regulating of mTORC1 signaling and inducing autophagy in Gln-dependent lung squamous cell carcinoma cell lines. Targeting glutaminolysis may represent a potential therapeutic strategy for the treatment of Gln-dependent lung squamous cell carcinomas.
Sigma-Aldrich
MISSION® esiRNA, targeting human GLS2