MilliporeSigma
  • Direct sample introduction-gas chromatography-mass spectrometry for the determination of haloanisole compounds in cork stoppers.
Direct sample introduction-gas chromatography-mass spectrometry for the determination of haloanisole compounds in cork stoppers.
Journal of chromatography. A (2016-11-15)
J I Cacho, J Nicolás, P Viñas, N Campillo, M Hernández-Córdoba
ABSTRACT
A solventless analytical method is proposed for analyzing the compounds responsible for cork taint in cork stoppers. Direct sample introduction (DSI) is evaluated as a sample introduction system for the gas chromatography-mass spectrometry (GC-MS) determination of four haloanisoles (HAs) in cork samples. Several parameters affecting the DSI step, including desorption temperature and time, gas flow rate and other focusing parameters, were optimized using univariate and multivariate approaches. The proposed method shows high sensitivity and minimises sample handling, with detection limits of 1.6-2.6ngg
Sigma-Aldrich
5-Bromo-2-chloroanisole, 97%