MilliporeSigma
  • Ovine plasma prion protein levels show genotypic variation detected by C-terminal epitopes not exposed in cell-surface PrPC.
Ovine plasma prion protein levels show genotypic variation detected by C-terminal epitopes not exposed in cell-surface PrPC.
The Biochemical journal (2006-08-03)
Alana M Thackray, Tim J Fitzmaurice, Lee Hopkins, Raymond Bujdoso
ABSTRACT
Ovine PBMCs (peripheral blood mononuclear cells) express PrP(C) [cellular PrP (prion-related protein)] and have the potential to harbour and release disease-associated forms of PrP during scrapie in sheep. Cell-surface PrP(C) expression by PBMCs, together with plasma PrP(C) levels, may contribute to the regulatory mechanisms that determine susceptibility and resistance to natural scrapie in sheep. Here, we have correlated cell-surface PrP(C) expression on normal ovine PBMCs by FACS with the presence of PrP(C) in plasma measured by capture-detector immunoassay. FACS showed similar levels of cell-surface PrP(C) on homozygous ARR (Ala136-Arg154-Arg171), ARQ (Ala136-Arg154-Gln171) and VRQ (Val136-Arg154-Gln171) PBMCs. Cell-surface ovine PrP(C) showed modulation of N-terminal epitopes, which was more evident on homozygous ARR cells. Ovine plasma PrP(C) levels showed genotypic variation and the protein displayed C-terminal epitopes not available in cell-surface PrP(C). Homozygous VRQ sheep showed the highest plasma PrP(C) level and homozygous ARR animals the lowest. For comparison, similar analyses were performed on normal bovine PBMCs and plasma. PrP(C) levels in bovine plasma were approx. 4-fold higher than ovine homozygous ARQ plasma despite similar levels of PBMC cell-surface PrP(C) expression. Immunoassays using C-terminal-specific anti-PrP monoclonal antibodies as capture and detector reagents revealed the highest level of PrP(C) in both ovine and bovine plasma, whilst lower levels were detected using N-terminal-specific monoclonal antibody FH11 as the capture reagent. This suggested that a proportion of plasma PrP(C) was N-terminally truncated. Our results indicate that the increased susceptibility to natural scrapie displayed by homozygous VRQ sheep correlates with a higher level of plasma PrP(C).
Sigma-Aldrich
Avidin–Alkaline Phosphatase, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution