MilliporeSigma
  • Extending colonic mucosal microbiome analysis-assessment of colonic lavage as a proxy for endoscopic colonic biopsies.
Extending colonic mucosal microbiome analysis-assessment of colonic lavage as a proxy for endoscopic colonic biopsies.
Microbiome (2016-11-26)
Euan Watt, Matthew R Gemmell, Susan Berry, Mark Glaire, Freda Farquharson, Petra Louis, Graeme I Murray, Emad El-Omar, Georgina L Hold
ABSTRACT
Sequencing-based analysis has become a well-established approach to deciphering the composition of the gut microbiota. However, due to the complexity of accessing sufficient material from colonoscopic biopsy samples, most studies have focused on faecal microbiota analysis, even though it is recognised that differences exist between the microbial composition of colonic biopsies and faecal samples. We determined the suitability of colonic lavage samples to see if it had comparable microbial diversity composition to colonic biopsies as they are without the limitations associated with sample size. We collected paired colonic biopsies and lavage samples from subjects who were attending for colorectal cancer screening colonoscopy. Next-generation sequencing and qPCR validation were performed with multiple bioinformatics analyses to determine the composition and predict function of the microbiota. Colonic lavage samples contained significantly higher numbers of operational taxonomic units (OTUs) compared to corresponding biopsy samples, however, diversity and evenness between lavage and biopsy samples were similar. The differences seen were driven by the presence of 12 OTUs which were in higher relative abundance in biopsies and were either not present or in low relative abundance in lavage samples, whilst a further 3 OTUs were present in higher amounts in the lavage samples compared to biopsy samples. However, predicted functional community profiling based on 16S ribosomal ribonucleic acid (rRNA) data indicated minimal differences between sample types. We propose that colonic lavage samples provide a relatively accurate representation of biopsy microbiota composition and should be considered where biopsy size is an issue.
Sigma-Aldrich
Magnesium citrate, AldrichCPR