MilliporeSigma
  • MafG controls the hypoxic response of cells by accumulating HIF-1alpha in the nuclei.
MafG controls the hypoxic response of cells by accumulating HIF-1alpha in the nuclei.
FEBS letters (2008-06-10)
Koji Ueda, Jing Xu, Haruka Morimoto, Atsumi Kawabe, Susumu Imaoka
ABSTRACT
We identified MafG as a protein that interacts with HIF-1alpha, a key factor in hypoxic response, using the yeast two-hybrid system. Interaction between MafG and HIF-1alpha was confirmed by surface plasmon resonance and by translocation to the nucleolus with the NoLS signal. A knockdown of MafG reduced erythropoietin (EPO) mRNA levels as well as luciferase reporter activity with the hypoxia response element. The knockdown of MafG did not change total HIF-1alpha protein, but reduced the accumulation of HIF-1alpha in the nuclei. These results suggest that MafG regulates the hypoxic response of cells by detaining HIF-1alpha in the nuclei.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid