MilliporeSigma
  • Activation-induced cytidine deaminase selectively catalyzed active DNA demethylation in pluripotency gene and improved cell reprogramming in bovine SCNT embryo.
Activation-induced cytidine deaminase selectively catalyzed active DNA demethylation in pluripotency gene and improved cell reprogramming in bovine SCNT embryo.
Cytotechnology (2016-08-11)
Xudong Ao, Rula Sa, Jie Wang, Rinuo Dao, Huimin Wang, Haiquan Yu
ABSTRACT
DNA methylation in mammals is an epigenetic marker and necessary for normal embryogenesis. The global genomic demethylation of 5-methylcytosine occurs during the first cell cycle following fertilization. Activation-induced cytidine deaminase (AID), which is well-known for the function in antibody diversification, has been implicated to play a role in active demethylation, but its role in cell reprogramming and its crosstalk with other DNA demethylation mechanism need to be clarified. In this study, the dynamic epigenetic regulation of cell pluripotency and embryo development by AID in bovine preimplantation embryos was investigated. The analysis of an AID overexpressing transgenic cell line showed that AID overexpression did not change the global genomic methylation but did change the methylation status of the promoters of the OCT4, NANOG and SOX2 genes, thereby causing changes in their expression. The siRNA-mediated AID knockdown in early embryonic development indicated that AID interference did not affect oocyte maturation or the following embryo development after in vitro fertilization but influenced the DNA methylation status of OCT4 and NANOG. To clarify the role of AID in preimplantation embryos, SCNT embryos were obtained using AID-overexpressing cells as nuclear donors. Compared to the control group, the cleavage and blastocyst rates were both significantly improved in the AID-overexpression group. The expression of OCT4 and NANOG was increased in the SCNT embryos, whereas the methylation levels of their promoters were reduced. In conclusion, this study demonstrated that AID selectively catalyzes DNA demethylation of pluripotency genes to play a role in regulation their expression, improves bovine SCNT embryo development by increased expression levels.
Sigma-Aldrich
MISSION® esiRNA, targeting human AICDA