MilliporeSigma
  • Pulmonary α-1,3-Glucan-Specific IgA-Secreting B Cells Suppress the Development of Cockroach Allergy.
Pulmonary α-1,3-Glucan-Specific IgA-Secreting B Cells Suppress the Development of Cockroach Allergy.
Journal of immunology (Baltimore, Md. : 1950) (2016-09-02)
Preeyam S Patel, R Glenn King, John F Kearney
ABSTRACT
There is a higher incidence of allergic conditions among children living in industrialized countries than those in developing regions. One explanation for this is reduced neonatal exposure to microbes and the consequent lack of immune stimulation. Sensitivity to cockroach allergen is highly correlated with the development of severe asthma. In this study, we determined that an Ab to microbial α-1,3-glucan binds an Enterobacter species and cockroach allergen. Neonatal, but not adult, mice immunized with this α-1,3-glucan-bearing Enterobacter (MK7) are protected against cockroach allergy. Following exposure to cockroach allergen, α-1,3-glucan-specific IgA-secreting cells are present in the lungs of mice immunized with MK7 as neonates but not in the lungs of those immunized as adults. Mice that are unable to generate anti-α-1,3-glucan IgA Abs were immunized with MK7 as neonates and were no longer protected against cockroach allergy. Thus, neonatal, but not adult, exposure to α-1,3-glucan results in suppressed development of cockroach allergy via pulmonary α-1,3-glucan-specific IgA-secreting cells.
Product Description
Latex beads, polystyrene, 3.0 μm mean particle size
Sigma-Aldrich
Triton X-405 solution, 70% in H2O