MilliporeSigma
  • Crosslinked polyarylene ether nitrile film as flexible dielectric materials with ultrahigh thermal stability.
Crosslinked polyarylene ether nitrile film as flexible dielectric materials with ultrahigh thermal stability.
Scientific reports (2016-11-09)
Ruiqi Yang, Renbo Wei, Kui Li, Lifen Tong, Kun Jia, Xiaobo Liu
ABSTRACT
Dielectric film with ultrahigh thermal stability based on crosslinked polyarylene ether nitrile is prepared and characterized. The film is obtained by solution-casting of polyarylene ether nitrile terminated phthalonitrile (PEN-Ph) combined with post self-crosslinking at high temperature. The film shows a 5% decomposition temperature over 520 °C and a glass transition temperature (T
Sigma-Aldrich
4-Nitrophthalonitrile, 99%