MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 8-Aminoguanosine and 8-Aminoguanine Exert Diuretic, Natriuretic, Glucosuric, and Antihypertensive Activity.
8-Aminoguanosine and 8-Aminoguanine Exert Diuretic, Natriuretic, Glucosuric, and Antihypertensive Activity.
The Journal of pharmacology and experimental therapeutics (2016-09-30)
Edwin K Jackson, Delbert G Gillespie, Zaichuan Mi
ABSTRACT
In vivo, guanine moieties in DNA, RNA, guanine nucleotides, or guanosine or guanine per se can undergo nitration (for example, by peroxynitrite) or hydroxylation (for example, by superoxide anion) on position 8 of the purine ring. Subsequent catabolism of these modified biomolecules leads to the production of a diverse group of 8-nitro, 8-amino, and 8-hydroxy guanosine and guanine compounds. Indeed, studies suggest the in vivo existence of 8-nitroguanosine, 8-nitroguanine, 8-aminoguanosine, 8-aminoguanine, 8-hydroxyguanosine, 8-hydroxy-2'-deoxyguanosine, and 8-hydroxyguanine. Since a multitude of these compounds exist in vivo, and since the renal effects of 8-substituted guanosine and guanine compounds are entirely unknown, we examined the effects of guanosine, guanine, 8-nitroguanosine, 8-nitroguanine, 8-hydroxyguanosine, 8-hydroxyguanine, 8-hydroxy-2'-deoxyguanosine, 8-aminoguanosine, and 8-aminoguanine (33.5 µmol/kg/min; intravenous infusion for 115 minutes) on excretion of sodium, potassium, and glucose in rats. Guanosine, 8-nitroguanosine, and 8-hydroxy-2'-deoxyguanosine had minimal natriuretic activity. Guanine, 8-nitroguanine, 8-hydroxyguanosine, and 8-hydroxyguanine had moderate natriuretic activity (increased sodium excretion by 9.4-, 7.8-, 7.1-, and 8.6-fold, respectively). In comparison with all other compounds, 8-aminoguanosine and 8-aminoguanine were highly efficacious and increased sodium excretion by 26.6- and 17.2-fold, respectively, exceeding that of a matched dose of amiloride (13.6-fold increase). 8-Aminoguanosine and 8-aminoguanine also increased glucose excretion by 12.1- and 12.2-fold, respectively, and decreased potassium excretion by 69.1 and 71.0%, respectively. Long-term radiotelemetry studies demonstrated that oral 8-aminoguanosine and 8-aminoguanine (5 mg/kg/day) suppressed deoxycorticosterone/salt-induced hypertension. These experiments demonstrate that some naturally occurring 8-substitued guanosine and guanine compounds, particularly 8-aminoguanosine and 8-aminoguanine, are potent and efficacious potassium-sparing diuretics/natriuretics that may represent a novel class of antihypertensive diuretics.
Sigma-Aldrich
8-Aminoguanosine, ≥98% (HPLC)
Sigma-Aldrich
8-Aminoguanine, ≥95% (HPLC)