MilliporeSigma
  • Real-time monitoring of specific oxygen uptake rates of embryonic stem cells in a microfluidic cell culture device.
Real-time monitoring of specific oxygen uptake rates of embryonic stem cells in a microfluidic cell culture device.
Biotechnology journal (2016-05-24)
Alexandre Super, Nicolas Jaccard, Marco Paulo Cardoso Marques, Rhys Jarred Macown, Lewis Donald Griffin, Farlan Singh Veraitch, Nicolas Szita
ABSTRACT
Oxygen plays a key role in stem cell biology as a signaling molecule and as an indicator of cell energy metabolism. Quantification of cellular oxygen kinetics, i.e. the determination of specific oxygen uptake rates (sOURs), is routinely used to understand metabolic shifts. However current methods to determine sOUR in adherent cell cultures rely on cell sampling, which impacts on cellular phenotype. We present real-time monitoring of cell growth from phase contrast microscopy images, and of respiration using optical sensors for dissolved oxygen. Time-course data for bulk and peri-cellular oxygen concentrations obtained for Chinese hamster ovary (CHO) and mouse embryonic stem cell (mESCs) cultures successfully demonstrated this non-invasive and label-free approach. Additionally, we confirmed non-invasive detection of cellular responses to rapidly changing culture conditions by exposing the cells to mitochondrial inhibiting and uncoupling agents. For the CHO and mESCs, sOUR values between 8 and 60 amol cell(-1) s(-1) , and 5 and 35 amol cell(-1) s(-1) were obtained, respectively. These values compare favorably with literature data. The capability to monitor oxygen tensions, cell growth, and sOUR, of adherent stem cell cultures, non-invasively and in real time, will be of significant benefit for future studies in stem cell biology and stem cell-based therapies.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture