MilliporeSigma
  • Physiological role of AMP-activated protein kinase (AMPK): insights from knockout mouse models.
Physiological role of AMP-activated protein kinase (AMPK): insights from knockout mouse models.
Biochemical Society transactions (2003-01-28)
B Viollet, F Andreelli, S B Jørgensen, C Perrin, D Flamez, J Mu, J F P Wojtaszewski, F C Schuit, M Birnbaum, E Richter, R Burcelin, S Vaulont
ABSTRACT
AMP-activated protein kinase (AMPK) is viewed as a fuel sensor for glucose and lipid metabolism. To understand better the physiological role of the catalytic AMPK subunit isoforms, we generated two knockout mouse models with the alpha1 (AMPK alpha 1(-/-)) and alpha 2 (AMPK alpha 2(-/-)) catalytic subunit genes deleted. No defect in glucose homoeostasis was observed in AMPK alpha 1(-/-) mice. On the other hand, AMPK alpha 2(-/-) mice presented high plasma glucose levels and low plasma insulin concentrations in the fed period and during the glucose tolerance test. Nevertheless, in isolated AMPK alpha 2(-/-) pancreatic islets, glucose-stimulated insulin secretion was not affected. Surprisingly, AMPK alpha 2(-/-) mice were insulin-resistant and had reduced muscle glycogen synthesis as assessed in vivo by the hyperinsulinaemic euglycaemic clamp procedure. Reduction of insulin sensitivity and glycogen synthesis were not dependent on the lack of AMPK in skeletal muscle, since mice expressing a dominant inhibitory mutant of AMPK in skeletal muscle were not affected and since insulin-stimulated glucose transport in incubated muscles in vitro was normal in AMPK alpha 2(-/-) muscles. Furthermore, AMPK alpha 2(-/-) mice have a higher sympathetic tone, as shown by increased catecholamine urinary excretion. Increased adrenergic tone could explain both decreased insulin secretion and insulin resistance observed in vivo in AMPK alpha 2(-/-) mice. We suggest that the alpha2 catalytic subunit of AMPK plays a major role as a fuel sensor by modulating the activity of the autonomous nervous system in vivo.
Sigma-Aldrich
AMPK (α1/β1/γ3), active, His tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
AMPK (α1/β2/γ1), active, His tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution