MilliporeSigma
  • Opposing roles of serine/threonine kinases MEKK1 and LOK in regulating the CD28 responsive element in T-cells.
Opposing roles of serine/threonine kinases MEKK1 and LOK in regulating the CD28 responsive element in T-cells.
The Biochemical journal (2002-03-21)
Li Tao, Scott Wadsworth, Jason Mercer, Cynthia Mueller, Kirsten Lynn, John Siekierka, Avery August
ABSTRACT
T-cell activation requires signals from both the T-cell receptor (TcR) and other co-stimulatory molecules such as CD28. TcR- and CD28-mediated signals are integrated during T-cell activation resulting in the expression of cytokine genes such as interleukin-2 (IL-2). An enhancer element (CD28RE) of the IL-2 gene specifically responsive to CD28 signals has been previously identified and characterized. This response element and an adjacent Activated Protein-1 (nuclear factor-interleukin-2B) site together (RE/AP1) were shown to complex with c-rel, AP-1 and other factors. However, details of the signal transduction pathways leading from CD28 to the composite response element remain poorly understood. We present data showing that overexpression of the serine threonine kinase, mitogen-activated protein kinase/extracellular-signal-regulated kinase kinase kinase-1 (MEKK1), but not nuclear factor-kappa B inducing kinase, or MAP kinase/ERK kinase-1 (MEK1), can significantly increase the level of CD28RE/AP1-driven luciferase (Luc) reporter gene expression in Jurkat E6-1 cells. A MEKK1 dominant negative mutant blocked such activation induced by stimulation with Raji B cells and the superantigen staphylococcus enterotoxin E (SEE), as well as via CD3/CD28. Mutations in either site of the RE/AP1 element abolished MEKK1-induced Luc expression. Calcineurin inhibitors, CsA and FK520, or inhibitors of p38 kinase (SB 203580), or MEK1 (PD 098059), did not affect MEKK1-induced reporter activation. These results directly implicate MEKK1 in the CD28 signalling pathway that activates the CD28 response element. Co-expression of the lymphocyte-oriented kinase (LOK) kinase attenuated Raji/SEE-induced IL-2 production in Jurkat cells, as well as MEKK1 and Raji/SEE-induced reporter gene activation. These data suggest that MEKK1 and LOK may have opposing roles in regulating the CD28RE/AP1 element.
Sigma-Aldrich
LOK (1-348), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution