MilliporeSigma
  • Identification and characterization of novel components of a Ca2+/calmodulin-dependent protein kinase cascade in HeLa cells.
Identification and characterization of novel components of a Ca2+/calmodulin-dependent protein kinase cascade in HeLa cells.
FEBS letters (2003-08-26)
Yumi Ishikawa, Hiroshi Tokumitsu, Hiroyuki Inuzuka, Maki Murata-Hori, Hiroshi Hosoya, Ryoji Kobayashi
ABSTRACT
In this report, we cloned a novel calmodulin-kinase (CaM-KIdelta) from HeLa cells and characterized its activation mechanism. CaM-KIdelta exhibits Ca(2+)/CaM-dependent activity that is enhanced (approximately 30-fold) in vitro by phosphorylation of its Thr180 by CaM-K kinase (CaM-KK)alpha, consistent with detection of CaM-KIdelta-activating activity in HeLa cells. We also identified a novel CaM-KKbeta isoform (CaM-KKbeta-3) in HeLa cells whose activity was highly Ca(2+)/CaM-independent. Transiently expressed CaM-KIdelta exhibited enhanced protein kinase activity in HeLa cells without ionomycin stimulation. This sustained activation of CaM-KIdelta was completely abolished by Thr180Ala mutation and inhibited by CaM-KK inhibitor, STO-609, indicating a functional CaM-KK/CaM-KIdelta cascade in HeLa cells.
Sigma-Aldrich
CAMK1δ, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution