MilliporeSigma
  • Increased Activity of Src Homology 2 Domain Containing Phosphotyrosine Phosphatase 2 (Shp2) Regulates Activity-dependent AMPA Receptor Trafficking.
Increased Activity of Src Homology 2 Domain Containing Phosphotyrosine Phosphatase 2 (Shp2) Regulates Activity-dependent AMPA Receptor Trafficking.
The Journal of biological chemistry (2016-07-16)
Bin Zhang, Yong-Lan Du, Wen Lu, Xun-Yi Yan, Qian Yang, Wei Yang, Jian-Hong Luo
ABSTRACT
Long term synaptic plasticity, such as long term potentiation (LTP), has been widely accepted as a cellular mechanism underlying memory. Recently, it has been unraveled that Shp2 plays a role in synaptic plasticity and memory in Drosophila and mice, revealing significant and conserved effects of Shp2 in cognitive function. However, the exact mechanism underlying this function of Shp2 in synaptic plasticity and memory still remains elusive. Here, we examine the regulation of Shp2 in hippocampal LTP and contextual fear conditioning. We find that Shp2 is rapidly recruited into spines after LTP induction. Furthermore, the phosphorylation level of Shp2 at Tyr-542 is elevated after LTP stimuli either in cultured hippocampal neurons or acute slices. Notably, contextual fear conditioning also regulates the phosphorylation level of Shp2 at Tyr-542, suggesting fine-tuned regulation of Shp2 in LTP and memory formation. By using a Shp2-specific inhibitor and adeno-associated virus-Cre mediated Shp2 knock-out in cultured neurons, we provide evidence that the phosphatase activity of Shp2 is critical for activity-dependent AMPA receptor surface trafficking. Collectively, our results have revealed a regulatory mechanism of Shp2 underlying LTP and memory, broadening our understanding of Shp2 in cognitive function.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GluR1-NT (NT) Antibody, clone RH95, clone RH95, from mouse