MilliporeSigma
  • Up-regulation of miR-455-5p by the TGF-β-SMAD signalling axis promotes the proliferation of oral squamous cancer cells by targeting UBE2B.
Up-regulation of miR-455-5p by the TGF-β-SMAD signalling axis promotes the proliferation of oral squamous cancer cells by targeting UBE2B.
The Journal of pathology (2016-05-29)
Chao-Min Cheng, Shine-Gwo Shiah, Chien-Chang Huang, Jenn-Ren Hsiao, Jang-Yang Chang
ABSTRACT
MicroRNAs (miRNAs) are involved in the tumourigenesis of various cancers by regulating their downstream targets. To identify the changes of miRNAs in oral squamous cell carcinoma (OSCC), we investigated the expression profiles of miRNAs in 40 pairs of OSCC specimens and their matched non-tumour epithelial tissues. Our data revealed higher miR-455-5p expression in the tumour tissues than in the normal tissues; the expression was also higher in oral cancer cell lines than in normal keratinocyte cell lines. MiR-455-5p knockdown reduced both the anchorage-independent growth and the proliferative ability of oral cancer cells, and these factors increased in miR-455-5p-overexpressing cells. Furthermore, by analysing the array data of patients with cancer and cell lines, we identified ubiquitin-conjugating enzyme E2B (UBE2B) as a target of miR-455-5p, and further validated this using 3'-untranslated region luciferase reporter assays and western blot analysis. We also demonstrated that UBE2B suppression rescued the impaired growth ability of miR-455-5p-knockdown cells. Furthermore, we observed that miR-455-5p expression was regulated, at least in part, by the transforming growth factor-β (TGF-β) pathway through the binding of SMAD3 to specific promoter regions. Notably, miR-455-5p expression was associated with the nodal status, stage, and overall survival in our patients, suggesting that miR-455-5p is a potential marker for predicting the prognosis of patients with oral cancer. In conclusion, we reveal that miR-455-5p expression is regulated by the TGF-β-dependent pathway, which subsequently leads to UBE2B down-regulation and contributes to oral cancer tumourigenesis. Copyright © 2016 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.
Product Description
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
TGF-β RI Kinase Inhibitor VI, SB431542, InSolution, ≥97%