MilliporeSigma
  • Histone Demethylation and Toll-like Receptor 8-Dependent Cross-Talk in Monocytes Promotes Transdifferentiation of Fibroblasts in Systemic Sclerosis Via Fra-2.
Histone Demethylation and Toll-like Receptor 8-Dependent Cross-Talk in Monocytes Promotes Transdifferentiation of Fibroblasts in Systemic Sclerosis Via Fra-2.
Arthritis & rheumatology (Hoboken, N.J.) (2016-01-28)
Marzena Ciechomska, Steven O'Reilly, Stefan Przyborski, Fiona Oakley, Katarzyna Bogunia-Kubik, Jacob M van Laar
ABSTRACT
To investigate whether epigenetic changes can modulate monocytes to produce tissue inhibitor of metalloproteinases 1 (TIMP-1) via Fra-2 (an activator protein 1 [AP-1] family member), a novel downstream mediator that promotes fibrogenesis. AP-1 transcription factors and TIMP-1 expression were measured in monocytes from systemic sclerosis (SSc) patients and healthy controls. Involvement of Fra-2 in the regulation of TIMP-1 following treatment with Toll-like receptor 8 (TLR-8) agonist was investigated using a luciferase activity assay and chromatin immunoprecipitation (ChIP) analysis. Expression of TIMP-1 and Fra-2 was determined in response to TLR-8 treatment and to different histone modifications, including 3'-deazaneplanocin (DZNep) and apicidin. Fibroblasts from healthy controls were cocultured with DZNep plus TLR-8-treated healthy control monocytes. Up-regulation of Fra-2 was detected in bleomycin-challenged mice and in skin biopsy samples from SSc patients. Enhanced expression of Fra-2 and TIMP-1 was correlated in SSc monocytes (P = 0.021). The expression of Fra-1 was significantly reduced (P = 0.037) in SSc monocytes. Inhibiting AP-1 activity reduced TIMP-1 production in TLR-8-stimulated monocytes from healthy controls and SSc patients. ChIP experiments revealed binding of Fra-2 to the TIMP-1 promoter. Stimulation with DZNep plus TLR-8 enhanced Fra-2 and TIMP-1 expression in healthy control monocytes, whereas TLR-8 plus apicidin repressed Fra-2 and TIMP-1 expression. Finally, healthy control monocytes treated with DZNep plus TLR-8 induced strong production of α-smooth muscle actin in dermal fibroblasts, which was inhibited by TIMP-1-blocking antibody. These data demonstrate a novel role of histone demethylation induced by DZNep on Fra-2-mediated TIMP-1 production by monocytes in the presence of TLR-8 agonist. This consequently orchestrates the transdifferentiation of fibroblasts, a key event in the pathogenesis of SSc.
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.