MilliporeSigma
  • A low-dose β1-blocker in combination with milrinone improves intracellular Ca2+ handling in failing cardiomyocytes by inhibition of milrinone-induced diastolic Ca2+ leakage from the sarcoplasmic reticulum.
A low-dose β1-blocker in combination with milrinone improves intracellular Ca2+ handling in failing cardiomyocytes by inhibition of milrinone-induced diastolic Ca2+ leakage from the sarcoplasmic reticulum.
PloS one (2015-01-24)
Shigeki Kobayashi, Takehisa Susa, Hironori Ishiguchi, Takeki Myoren, Wakako Murakami, Takayoshi Kato, Masakazu Fukuda, Akihiro Hino, Takeshi Suetomi, Makoto Ono, Hitoshi Uchinoumi, Hiroki Tateishi, Mamoru Mochizuki, Tetsuro Oda, Shinichi Okuda, Masahiro Doi, Takeshi Yamamoto, Masafumi Yano
ABSTRACT
The purpose of this study was to investigate whether adding a low-dose β1-blocker to milrinone improves cardiac function in failing cardiomyocytes and the underlying cardioprotective mechanism. The molecular mechanism underlying how the combination of low-dose β1-blocker and milrinone affects intracellular Ca(2+) handling in heart failure remains unclear. We investigated the effect of milrinone plus landiolol on intracellular Ca(2+) transient (CaT), cell shortening (CS), the frequency of diastolic Ca(2+) sparks (CaSF), and sarcoplasmic reticulum Ca(2+) concentration ({Ca(2+)}SR) in normal and failing canine cardiomyocytes and used immunoblotting to determine the phosphorylation level of ryanodine receptor (RyR2) and phospholamban (PLB). In failing cardiomyocytes, CaSF significantly increased, and peak CaT and CS markedly decreased compared with normal myocytes. Administration of milrinone alone slightly increased peak CaT and CS, while CaSF greatly increased with a slight increase in {Ca(2+)}SR. Co-administration of β1-blocker landiolol to failing cardiomyocytes at a dose that does not inhibit cardiomyocyte function significantly decreased CaSF with a further increase in {Ca(2+)}SR, and peak CaT and CS improved compared with milrinone alone. Landiolol suppressed the hyperphosphorylation of RyR2 (Ser2808) in failing cardiomyocytes but had no effect on levels of phosphorylated PLB (Ser16 and Thr17). Low-dose landiolol significantly inhibited the alternans of CaT and CS under a fixed pacing rate (0.5 Hz) in failing cardiomyocytes. A low-dose β1-blocker in combination with milrinone improved cardiac function in failing cardiomyocytes, apparently by inhibiting the phosphorylation of RyR2, not PLB, and subsequent diastolic Ca(2+) leak.
Sigma-Aldrich
Landiolol hydrochloride, ≥98% (HPLC)