MilliporeSigma
Nigericin-induced death of an acidophilic bacterium.
Journal of general microbiology (1979-09-01)
A A Guffanti, L F Davidson, T M Mann, T A Krulwich
PMID42667
ABSTRACT
At an external pH of 3.5, nigericin (which catalyses an electroneutral H+/K+ exchange) abolished the transmembrane proton gradient (delta pH) of Bacillus acidocaldarius, causing a rapid acidification of the cytoplasm from approximately pH 6.0 to pH 3.5. A pronounced loss of viability and fine-structural changes rapidly followed treatment with nigericin. A marked decline in respiration and an even more rapid decrease in cytoplasmic ATP were observed. Activity of at least one cytoplasmic enzyme decreased more slowly. There was no generalized loss in the integrity of the cytoplasmic membrane, as assayed by permeability to inulin or Na+ or by release of ultraviolet light-absorbing compounds. The loss of viability upon treatment with carbonyl cyanide m-chlorophenylhydrazone was similar to what observed with nigericin, so proton influx alone, rather than together with K+ efflux, was probably involved in the death of the organism. Moreover, acidification of the cytoplasm rather than abolition of the delta pH was the lethal event, since no loss of viability was observed when the delta pH was abolished by elevation of the external pH.
Sigma-Aldrich
Nigericin sodium salt Ready Made Solution, 5 mg/mL (DMSO:ethanol 1:1)
Pricing and availability is not currently available.