MilliporeSigma
  • Efficient synthesis of an adenosine A2a agonist: glycosylation of 2-haloadenines and an N2-alkyl-6-chloroguanine.
Efficient synthesis of an adenosine A2a agonist: glycosylation of 2-haloadenines and an N2-alkyl-6-chloroguanine.
The Journal of organic chemistry (2004-04-24)
John M Caddell, Alan M Chapman, Bob E Cooley, Brian P Downey, Michael P LeBlanc, Mary M Jackson, Thomas M O'Connell, Hahn-My Phung, Thomas D Roper, Shiping Xie
ABSTRACT
A convergent synthesis of adenosine A2a agonist 1 in the form of its maleate salt 2 was achieved. The key step in this approach was the highly selective 9beta-glycosylation reaction between 2-haloadenines or an N(2)-alkyl-6-chloroguanine and a D-ribose derivative containing a 2-ethyltetrazolyl moiety. Glycosylations of other purine derivatives were also examined, and the methods developed provide efficient access to a variety of adenosine analogues such as 2-alkylaminoadenosines, an attractive class of compounds with antiinflammatory activity.
Sigma-Aldrich
2-Bromohypoxanthine, 96%