MilliporeSigma
Lrp4 in astrocytes modulates glutamatergic transmission.
Nature neuroscience (2016-06-14)
Xiang-Dong Sun, Lei Li, Fang Liu, Zhi-Hui Huang, Jonathan C Bean, Hui-Feng Jiao, Arnab Barik, Seon-Myung Kim, Haitao Wu, Chengyong Shen, Yun Tian, Thiri W Lin, Ryan Bates, Anupama Sathyamurthy, Yong-Jun Chen, Dong-Min Yin, Lei Xiong, Hui-Ping Lin, Jin-Xia Hu, Bao-Ming Li, Tian-Ming Gao, Wen-Cheng Xiong, Lin Mei
ABSTRACT
Neurotransmission requires precise control of neurotransmitter release from axon terminals. This process is regulated by glial cells; however, the underlying mechanisms are not fully understood. We found that glutamate release in the brain was impaired in mice lacking low-density lipoprotein receptor-related protein 4 (Lrp4), a protein that is critical for neuromuscular junction formation. Electrophysiological studies revealed compromised release probability in astrocyte-specific Lrp4 knockout mice. Lrp4 mutant astrocytes suppressed glutamatergic transmission by enhancing the release of ATP, whose level was elevated in the hippocampus of Lrp4 mutant mice. Consequently, the mutant mice were impaired in locomotor activity and spatial memory and were resistant to seizure induction. These impairments could be ameliorated by blocking the adenosine A1 receptor. The results reveal a critical role for Lrp4, in response to agrin, in modulating astrocytic ATP release and synaptic transmission. Our findings provide insight into the interaction between neurons and astrocytes for synaptic homeostasis and/or plasticity.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
EHNA hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-GluR2 Antibody, clone L21/32, clone L21/32, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Akt Inhibitor IV, The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.