MilliporeSigma
Intracellular insulin quantification by cell-ELISA.
Experimental cell research (2016-06-28)
Parker Lyng Andersen, Patrick Vermette
ABSTRACT
Current methods of monitoring insulin in culture are limited to soluble insulin (secretions or lysates) or synthetic gene reporter analyses. We present an insulin-specific cell enzyme-linked immunosorbent assay (cell-ELISA) to assess relative intracellular insulin protein content of adherent cultures, normalized to cell density with further immunocytochemical verification of insulin-expressing cells within identical cultures. The protocol was optimized and validated using an insulin-expressing cell line (INS-1) by confirming direct relations between intracellular insulin content and insulin-expressing cell density, in a glucose exposure-dependent manner. Utility was demonstrated by identifying multiple INS-1 clones lowly expressing insulin following lentiviral particle delivery of interference RNA intended to down-regulate one of the insulin gene-directed transcription factors, MafA. The cell-ELISA was also applied to monitor insulin content within partially dissociated primary mouse islet cultures. This technique allows for the first time routine analysis of intracellular insulin protein in vitro suitable for investigating islet cell biology by means of multiple parameter screening.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Janus Green B, certified by the Biological Stain Commission, Dye content 65 %