MilliporeSigma
  • Insulin and IGF-1 receptors regulate FoxO-mediated signaling in muscle proteostasis.
Insulin and IGF-1 receptors regulate FoxO-mediated signaling in muscle proteostasis.
The Journal of clinical investigation (2016-08-16)
Brian T O'Neill, Kevin Y Lee, Katherine Klaus, Samir Softic, Megan T Krumpoch, Joachim Fentz, Kristin I Stanford, Matthew M Robinson, Weikang Cai, Andre Kleinridders, Renata O Pereira, Michael F Hirshman, E Dale Abel, Domenico Accili, Laurie J Goodyear, K Sreekumaran Nair, C Ronald Kahn
ABSTRACT
Diabetes strongly impacts protein metabolism, particularly in skeletal muscle. Insulin and IGF-1 enhance muscle protein synthesis through their receptors, but the relative roles of each in muscle proteostasis have not been fully elucidated. Using mice with muscle-specific deletion of the insulin receptor (M-IR-/- mice), the IGF-1 receptor (M-IGF1R-/- mice), or both (MIGIRKO mice), we assessed the relative contributions of IR and IGF1R signaling to muscle proteostasis. In differentiated muscle, IR expression predominated over IGF1R expression, and correspondingly, M-IR-/- mice displayed a moderate reduction in muscle mass whereas M-IGF1R-/- mice did not. However, these receptors serve complementary roles, such that double-knockout MIGIRKO mice displayed a marked reduction in muscle mass that was linked to increases in proteasomal and autophagy-lysosomal degradation, accompanied by a high-protein-turnover state. Combined muscle-specific deletion of FoxO1, FoxO3, and FoxO4 in MIGIRKO mice reversed increased autophagy and completely rescued muscle mass without changing proteasomal activity. These data indicate that signaling via IR is more important than IGF1R in controlling proteostasis in differentiated muscle. Nonetheless, the overlap of IR and IGF1R signaling is critical to the regulation of muscle protein turnover, and this regulation depends on suppression of FoxO-regulated, autophagy-mediated protein degradation.
Sigma-Aldrich
Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin, protein C substrate
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Tamoxifen, ≥99%