MilliporeSigma
  • Optimal myelin elongation relies on YAP activation by axonal growth and inhibition by Crb3/Hippo pathway.
Optimal myelin elongation relies on YAP activation by axonal growth and inhibition by Crb3/Hippo pathway.
Nature communications (2016-07-21)
Ruani N Fernando, Laurent Cotter, Claire Perrin-Tricaud, Jade Berthelot, Sylvain Bartolami, Jorge A Pereira, Sergio Gonzalez, Ueli Suter, Nicolas Tricaud
ABSTRACT
Fast nerve conduction relies on successive myelin segments that electrically isolate axons. Segment geometry-diameter and length-is critical for the optimization of nerve conduction and the molecular mechanisms allowing this optimized geometry are partially known. We show here that peripheral myelin elongation is dynamically regulated by stimulation of YAP (Yes-associated protein) transcription cofactor activity during axonal elongation and limited by inhibition of YAP activity via the Hippo pathway. YAP promotes myelin and non-myelin genes transcription while the polarity protein Crb3, localized at the tips of the myelin sheath, activates the Hippo pathway to temper YAP activity, therefore allowing for optimal myelin growth. Dystrophic Dy(2j/2j) mice mimicking human peripheral neuropathy with reduced internodal lengths have decreased nuclear YAP which, when corrected, leads to longer internodes. These data show a novel mechanism controlling myelin growth and nerve conduction, and provide a molecular ground for disease with short myelin segments.
Sigma-Aldrich
Anti-AnkyrinG Mouse mAb (463), liquid, ≥95% (SDS-PAGE), clone 463
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture