MilliporeSigma
  • Ferulic acid and benzothiazole dimer derivatives with high binding affinity to beta-amyloid fibrils.
Ferulic acid and benzothiazole dimer derivatives with high binding affinity to beta-amyloid fibrils.
Bioorganic & medicinal chemistry letters (2007-06-05)
Seong Rim Byeon, Yun Jung Jin, Soo Jeong Lim, Ji Hoon Lee, Kyung Ho Yoo, Kye Jung Shin, Seung Jun Oh, Dong Jin Kim
ABSTRACT
New ferulic acid and benzothiazole dimer derivatives were synthesized and evaluated by in vitro competition assay using [(125)I]TZDM for their specific binding affinities to Abeta fibrils. In particular, 4a showed the most excellent binding affinity (K(i)=0.53 nM), compared to PIB (K(i)=0.77 nM), for benzothiazole binding sites of Abeta(1-42) fibrils. This result suggests a possibility of a potential AD diagnostic probe for detection of Abeta fibrils.
Product Description
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, 99%