MilliporeSigma
  • FGFR3 expression in primary and metastatic urothelial carcinoma of the bladder.
FGFR3 expression in primary and metastatic urothelial carcinoma of the bladder.
Cancer medicine (2014-05-23)
Elizabeth A Guancial, Lillian Werner, Joaquim Bellmunt, Aristotle Bamias, Toni K Choueiri, Robert Ross, Fabio A Schutz, Rachel S Park, Robert J O'Brien, Michelle S Hirsch, Justine A Barletta, David M Berman, Rosina Lis, Massimo Loda, Edward C Stack, Levi A Garraway, Markus Riester, Franziska Michor, Philip W Kantoff, Jonathan E Rosenberg
ABSTRACT
While fibroblast growth factor receptor 3 (FGFR3) is frequently mutated or overexpressed in nonmuscle-invasive urothelial carcinoma (UC), the prevalence of FGFR3 protein expression and mutation remains unknown in muscle-invasive disease. FGFR3 protein and mRNA expression, mutational status, and copy number variation were retrospectively analyzed in 231 patients with formalin-fixed paraffin-embedded primary UCs, 33 metastases, and 14 paired primary and metastatic tumors using the following methods: immunohistochemistry, NanoString nCounterTM, OncoMap or Affymetrix OncoScanTM array, and Gain and Loss of Analysis of DNA and Genomic Identification of Significant Targets in Cancer software. FGFR3 immunohistochemistry staining was present in 29% of primary UCs and 49% of metastases and did not impact overall survival (P = 0.89, primary tumors; P = 0.78, metastases). FGFR3 mutations were observed in 2% of primary tumors and 9% of metastases. Mutant tumors expressed higher levels of FGFR3 mRNA than wild-type tumors (P < 0.001). FGFR3 copy number gain and loss were rare events in primary and metastatic tumors (0.8% each; 3.0% and 12.3%, respectively). FGFR3 immunohistochemistry staining is present in one third of primary muscle-invasive UCs and half of metastases, while FGFR3 mutations and copy number changes are relatively uncommon.