MilliporeSigma
  • Gastrin upregulates the prosurvival factor secretory clusterin in adenocarcinoma cells and in oxyntic mucosa of hypergastrinemic rats.
Gastrin upregulates the prosurvival factor secretory clusterin in adenocarcinoma cells and in oxyntic mucosa of hypergastrinemic rats.
American journal of physiology. Gastrointestinal and liver physiology (2011-10-15)
Christina Sæten Fjeldbo, Ingunn Bakke, Sten Even Erlandsen, Jannicke Holmseth, Astrid Lægreid, Arne K Sandvik, Liv Thommesen, Torunn Bruland
ABSTRACT
We show that the gastric hormone gastrin induces the expression of the prosurvival secretory clusterin (sCLU) in rat adenocarcinoma cells. Clusterin mRNA was still upregulated in the presence of the protein synthesis inhibitor cycloheximide, although at a lower level. This indicates that gastrin induces clusterin transcription independently of de novo protein synthesis but requires de novo protein synthesis of signal transduction pathway components to achieve maximal expression level. Luciferase reporter assay indicates that the AP-1 transcription factor complex is involved in gastrin-mediated activation of the clusterin promoter. Gastrin-induced clusterin expression and subsequent secretion is dependent on sustained treatment, because removal of gastrin after 1-2 h abolished the response. Neutralization of secreted clusterin by a specific antibody abolished the antiapoptotic effect of gastrin on serum starvation-induced apoptosis, suggesting that extracellular clusterin is involved in gastrin-mediated inhibition of apoptosis. The clusterin response to gastrin was validated in vivo in hypergastrinemic rats, showing increased clusterin expression in the oxyntic mucosa, as well as higher levels of clusterin in plasma. In normal rat oxyntic mucosa, clusterin protein was strongly expressed in chromogranin A-immunoreactive neuroendocrine cells, of which the main cell type was the histidine decarboxylase-immunoreactive enterochromaffin-like (ECL) cell. The association of clusterin with neuroendocrine differentiation was further confirmed in human gastric ECL carcinoids. Interestingly, in hypergastrinemic rats, clusterin-immunoreactive cells formed distinct groups of diverse cells at the base of many glands. Our results suggest that clusterin may contribute to gastrin's growth-promoting effect on the oxyntic mucosa.