MilliporeSigma
  • Structure-activity relationships of new N-acylanthranilic acid derivatives as plasminogen activator inhibitor-1 inhibitors.
Structure-activity relationships of new N-acylanthranilic acid derivatives as plasminogen activator inhibitor-1 inhibitors.
Chemical & pharmaceutical bulletin (2011-02-08)
Nagahisa Yamaoka, Hidehiko Kodama, Yuko Izuhara, Toshio Miyata, Kanji Meguro
ABSTRACT
Novel anthranilic acid derivatives having substituted N-acyl side chains were designed and synthesized for evaluation as plasminogen activator inhibitor-1 (PAI-1) inhibitors. Compounds with a 4-diphenylmethyl-1-piperazinyl moiety on the acyl side chains in general exhibited potent in vitro PAI-1 inhibitory activity and good pharmacokinetic profiles after oral administration in rats. Compound 16f (TM5275) was identified as a promising candidate for further pharmacological evaluation.
Sigma-Aldrich
Methyl 2-amino-5-bromobenzoate, 96%
Sigma-Aldrich
3-Oxaspiro[5,5]undecane-2,4-dione, 98%