MilliporeSigma
  • Inhibition of Kallikrein-Related Peptidases 7 and 5 by Grafting Serpin Reactive-Center Loop Sequences onto Sunflower Trypsin Inhibitor-1 (SFTI-1).
Inhibition of Kallikrein-Related Peptidases 7 and 5 by Grafting Serpin Reactive-Center Loop Sequences onto Sunflower Trypsin Inhibitor-1 (SFTI-1).
Chembiochem : a European journal of chemical biology (2015-11-18)
Cathleen Jendrny, Annette G Beck-Sickinger
ABSTRACT
Serpin proteins irreversibly inhibit serine proteases, but only a small part of the serpin reactive-center loop (RCL) is responsible for the initial protein-protein interaction (PPI). To develop peptidic protease inhibitors, kallikrein-related peptidases 7 (KLK7) and 5 (KLK5) were chosen. Firstly, we demonstrated that short peptides derived from RCL sequences can be cleaved by KLK7 in a substrate-like manner. Next, these substrates were grafted onto the protease-binding loop of sunflower trypsin inhibitor-1 (SFTI-1). Peptides based on kallistatin, α1 -antichymotrypsin, and protein C inhibitor (PCI) inhibited KLK7 with Ki =0.4, 0.5, and 0.7 μm, respectively. In contrast, the trypsin-like KLK5 was only blocked by the peptide derived from PCI (Ki =0.6 μm). Thus, serpin function can be mimicked by introducing its PPI site into the rigid structure of the SFTI-1 scaffold. This approach might be applicable not only to KLKs but also to other serine protease members, thus opening up new therapeutic fields.
Sigma-Aldrich
Fmoc-Gln(Trt)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)