MilliporeSigma
  • Synthesis of CdTe nanocrystals using Te nanorods as the Te source and the formation of microtubes with red fluorescence.
Synthesis of CdTe nanocrystals using Te nanorods as the Te source and the formation of microtubes with red fluorescence.
Inorganic chemistry (2007-07-31)
Rongbo Zheng, Shaojun Guo, Shaojun Dong
ABSTRACT
Here, we report an alternative route to the preparation of highly luminescent CdTe nanocrystals (NCs) using Te nanorods instead of freshly prepared NaHTe as the Te source via a one-pot route under hydrothermal conditions. Furthermore, microtubes with red fluorescence were generated via simply aging the above CdTe NC suspension under ambient conditions.
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis