MilliporeSigma
  • In the absence of overt urothelial damage, chondroitinase ABC digestion of the GAG layer increases bladder permeability in ovariectomized female rats.
In the absence of overt urothelial damage, chondroitinase ABC digestion of the GAG layer increases bladder permeability in ovariectomized female rats.
American journal of physiology. Renal physiology (2016-02-26)
Robert E Hurst, Samuel Van Gordon, Karl Tyler, Bradley Kropp, Rheal Towner, HsuehKung Lin, John O Marentette, Jane McHowat, Ehsan Mohammedi, Beverley Greenwood-Van Meerveld
ABSTRACT
Loss of integrity of the protective impermeability barrier in the urothelium has been identified as significant in bladder dysfunction. In this study, we tested the theory that the luminal layer of glycosaminoglycans (GAG) serves as an important component of barrier function. The peptide polycation protamine sulfate (PS), 1 mg/ml, was instilled intravesically for 10 min into rat bladders. Chondroitinase ABC (ChABC), 63 IU/ml, was instilled into an additional six rats for 30 min to digest the GAG layer. Unmanipulated controls and sham-injected controls were also performed. After 24 h, the rats were euthanized, the bladders were removed, and permeability was assessed in the Ussing chamber and by diffusion of FITC-labeled dextran (4 kDa) to measure macromolecular permeability. The status of tight junctions was assessed by immunofluorescence and electron microscopy. In control and sham treated rat bladders, the transepithelial electrical resistance were means of 2.5 ± 1.1 vs. 2.6 ± 1.1 vs 1.2 ± 0.5 and 1.01 ± 0.7 kΩ·cm(2) in the PS-treated and ChABC-treated rat bladders (P = 0.0016 and P = 0.0039, respectively). Similar differences were seen in dextran permeability. Histopathology showed a mild inflammation following PS treatment, but the ChABC-treated bladders were indistinguishable from controls. Tight junctions generally remained intact. ChABC digestion alone induced bladder permeability, confirming the importance of the GAG layer to bladder barrier function and supports that loss of the GAG layer seen in bladder biopsies of interstitial cystitis patients could be a significant factor producing symptoms for at least some interstitial cystitis/painful bladder syndrome patients.
Sigma-Aldrich
Anti-Chondroitin 6 Sulfate Antibody, clone MK-302, ascites fluid, clone MK-302, Chemicon®
Pricing and availability is not currently available.