MilliporeSigma
  • Novel vanilloid receptor-1 antagonists: 1. Conformationally restricted analogues of trans-cinnamides.
Novel vanilloid receptor-1 antagonists: 1. Conformationally restricted analogues of trans-cinnamides.
Journal of medicinal chemistry (2007-06-26)
Mark H Norman, Jiawang Zhu, Christopher Fotsch, Yunxin Bo, Ning Chen, Partha Chakrabarti, Elizabeth M Doherty, Narender R Gavva, Nobuko Nishimura, Thomas Nixey, Vassil I Ognyanov, Robert M Rzasa, Markian Stec, Sekhar Surapaneni, Rami Tamir, Vellarkad N Viswanadhan, James J S Treanor
ABSTRACT
The vanilloid receptor-1 (VR1 or TRPV1) is a member of the transient receptor potential (TRP) family of ion channels and plays a role as an integrator of multiple pain-producing stimuli. From a high-throughput screening assay, measuring calcium uptake in TRPV1-expressing cells, we identified an N-aryl trans-cinnamide (AMG9810, compound 9) that acts as a potent TRPV1 antagonist. We have demonstrated the antihyperalgesic properties of 9 in vivo and have also reported the discovery of novel, orally bioavailable cinnamides derived from this lead. Herein, we expand our investigations and describe the synthesis and biological evaluation of a series of conformationally constrained analogues of the s-cis conformer of compound 9. These investigations resulted in the identification of 4-amino- and 4-oxopyrimidine cores as suitable isosteric replacements for the trans-acrylamide moiety. The best examples from this series, pyrimidines 79 and 74, were orally bioavailable and exhibited potent antagonism of both rat (IC50 = 4.5 and 0.6 nM, respectively) and human TRPV1 (IC50 = 7.4 and 3.7 nM, respectively). In addition, compound 74 was shown to be efficacious at blocking a TRPV1-mediated physiological response in vivo in the capsaicin-induced hypothermia model in rats; however, it was ineffective at preventing thermal hyperalgesia induced by complete Freund's adjuvant in rats.
Sigma-Aldrich
2,4-Dichloropyrimidine, 98%
Sigma-Aldrich
4-Chloro-2-methylthiopyrimidine, 98%