MilliporeSigma
  • Methamphetamine-like discriminative stimulus effects of bupropion and its two hydroxy metabolites in male rhesus monkeys.
Methamphetamine-like discriminative stimulus effects of bupropion and its two hydroxy metabolites in male rhesus monkeys.
Behavioural pharmacology (2016-02-18)
Matthew L Banks, Douglas A Smith, Bruce E Blough
ABSTRACT
The dopamine transporter (DAT) inhibitor and nicotinic acetylcholine (nACh) receptor antagonist bupropion is being investigated as a candidate 'agonist' medication for methamphetamine addiction. In addition to its complex pharmacology, bupropion also has two distinct pharmacologically active metabolites. However, the mechanism by which bupropion produces methamphetamine-like 'agonist' effects remains unknown. The aim of the present study was to determine the role of DAT inhibition, nACh receptor antagonism, and the hydroxybupropion metabolites in the methamphetamine-like discriminative stimulus effects of bupropion in rhesus monkeys. In addition, varenicline, a partial agonist at the nACh receptor, and risperidone, a dopamine antagonist, were tested as controls. Monkeys (n=4) were trained to discriminate 0.18 mg/kg intramuscular methamphetamine from saline in a two-key food-reinforced discrimination procedure. The potency and time course of methamphetamine-like discriminative stimulus effects were determined for all compounds. Bupropion, methylphenidate, and 2S,3S-hydroxybupropion produced full, at least 90%, methamphetamine-like effects. 2R,3R-Hydroxybupropion, mecamylamine, and nicotine also produced full methamphetamine-like effects, but drug potency was more variable between monkeys. Varenicline produced partial methamphetamine-like effects, whereas risperidone did not. Overall, these results suggest DAT inhibition as the major mechanism of the methamphetamine-like 'agonist' effects of bupropion, although nACh receptor antagonism appeared, at least partially, to contribute. Furthermore, the contribution of the 2S,3S-hydroxybupropion metabolite could not be completely ruled out.
Sigma-Aldrich
(2S,3S)-Hydroxybupropion hydrochloride, ≥98% (HPLC)