MilliporeSigma
  • Regio- and stereospecific synthesis of beta-sulfonamidodisulfides and beta-sulfonamidosulfides from aziridines using tetrathiomolybdate as a sulfur transfer reagent.
Regio- and stereospecific synthesis of beta-sulfonamidodisulfides and beta-sulfonamidosulfides from aziridines using tetrathiomolybdate as a sulfur transfer reagent.
The Journal of organic chemistry (2007-02-24)
Devarajulu Sureshkumar, Thanikachalam Gunasundari, Venkataraman Ganesh, Srinivasan Chandrasekaran
ABSTRACT
A comprehensive study of a general and effective one-step procedure for the synthesis of beta-sulfonamidodisulfides directly from N-tosyl aziridines in a regio- and stereospecific manner under neutral conditions without the use of any Lewis acid or base has been reported. This methodology is extended to the synthesis of an optically pure cyclic seven-membered disulfide 29. Synthesis of a variety of beta-sulfonamidosulfides involving tandem, multistep reactions in one pot is also reported.
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis