MilliporeSigma
  • Beta-aminoethyltrifluoroborates: efficient aminoethylations via Suzuki-Miyaura cross-coupling.
Beta-aminoethyltrifluoroborates: efficient aminoethylations via Suzuki-Miyaura cross-coupling.
Organic letters (2007-01-16)
Gary A Molander, Fabricio Vargas
ABSTRACT
A set of phenethylamines has been successfully prepared via Suzuki-Miyaura cross-coupling of diverse potassium beta-aminoethyltrifluoroborates with aryl halides. The potassium beta-aminoethyltrifluoroborates were easily prepared via hydroboration of enamine and enamide precursors. [reaction: see text].
Sigma-Aldrich
RuPhos, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
RuPhos ChemBeads
Pricing and availability is not currently available.