MilliporeSigma
  • Hepatic differentiation of human pluripotent stem cells on human liver progenitor HepaRG-derived acellular matrix.
Hepatic differentiation of human pluripotent stem cells on human liver progenitor HepaRG-derived acellular matrix.
Experimental cell research (2016-02-09)
Liisa K Kanninen, Pauliina Porola, Johanna Niklander, Melina M Malinen, Anne Corlu, Christiane Guguen-Guillouzo, Arto Urtti, Marjo L Yliperttula, Yan-Ru Lou
ABSTRACT
Human hepatocytes are extensively needed in drug discovery and development. Stem cell-derived hepatocytes are expected to be an improved and continuous model of human liver to study drug candidates. Generation of endoderm-derived hepatocytes from human pluripotent stem cells (hPSCs), including human embryonic stem cells and induced pluripotent stem cells, is a complex, challenging process requiring specific signals from soluble factors and insoluble matrices at each developmental stage. In this study, we used human liver progenitor HepaRG-derived acellular matrix (ACM) as a hepatic progenitor-specific matrix to induce hepatic commitment of hPSC-derived definitive endoderm (DE) cells. The DE cells showed much better attachment to the HepaRG ACM than other matrices tested and then differentiated towards hepatic cells, which expressed hepatocyte-specific makers. We demonstrate that Matrigel overlay induced hepatocyte phenotype and inhibited biliary epithelial differentiation in two hPSC lines studied. In conclusion, our study demonstrates that the HepaRG ACM, a hepatic progenitor-specific matrix, plays an important role in the hepatic differentiation of hPSCs.
Product Description
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Pricing and availability is not currently available.