MilliporeSigma
  • An air-stable P-chiral phosphine ligand for highly enantioselective transition-metal-catalyzed reactions.
An air-stable P-chiral phosphine ligand for highly enantioselective transition-metal-catalyzed reactions.
Journal of the American Chemical Society (2005-08-25)
Tsuneo Imamoto, Keitaro Sugita, Kazuhiro Yoshida
ABSTRACT
A new P-chiral phosphine ligand, (R,R)-2,3-bis(tert-butylmethylphosphino)quinoxaline, has been prepared by the reaction of enantiomerically pure tert-butylmethylphosphine-borane with 2,3-dichloroquinoxaline. This ligand, in contrast to most of the previously reported P-chiral ligands, is an air-stable solid and exhibits excellent enantioselectivities in both Rh-catalyzed asymmetric hydrogenations and Rh- or Pd-catalyzed carbon-carbon bond-forming reactions.
Sigma-Aldrich
(R,R)-(–)-2,3-Bis(tert-butylmethylphosphino)quinoxaline, ≥95%